Learn More
PSME4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18799-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is expected to recognize all 4 isoforms of PSME4 but appears to mainly detect isoform 2 of 140 kDa.
proteasome (prosome, macropain) activator subunit 4 (PSME4) is subunit of the proteasome. PSME4 has been found to activate proteasomal cleavage of peptides in an energy-independent manner and may be involved in spermatogenesis. PSME4 may also be involved in DNA repair.Specifications
PSME4 | |
Polyclonal | |
Unconjugated | |
PSME4 | |
KIAA0077, PA200, Proteasome activator PA200, PSME4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103554, 23198, 498433 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14997, Q5SSW2 | |
Psme4 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
PSME4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction