missing translation for 'onlineSavingsMsg'
Learn More

PTBP1 Mouse anti-Human, Unconjugated, Clone: 3D6D8, Proteintech

Mouse Monoclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen PTBP1
Clone 3D6D8
Concentration 1000 ÎĽg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16800106
View Documents
Proteintech
67462-1-IG-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16890096
View Documents
Proteintech
67462-1-IG-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The PTBP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTBP1. This antibody recognizes human, mouse, rat antigen. The PTBP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.

Specifications

Specifications

PTBP1
1000 ÎĽg/mL
Monoclonal
Mouse
5725
Primary
3D6D8
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Unconjugated
Human, Rat, Mouse
IgG1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PTBP1 Mouse anti-Human, Unconjugated, Clone: 3D6D8, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.