missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55186-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to PTBP2.
PTBP2 encodes a protein that binds to the intronic cluster of RNA regulatory elements, downstream control sequence (DCS). It is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein but it is expressed primarily in the brain.Specifications
PTBP2 | |
Polyclonal | |
Unconjugated | |
PTBP2 | |
brPTB, PTB, PTB like protein, PTBLP, PTBP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
310820, 56195, 58155 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q66H20, Q91Z31, Q9UKA9 | |
Ptbp2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |