missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PTBP2 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836675
|
Proteintech
55186-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826675
|
Proteintech
55186-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to PTBP2.
PTBP2 encodes a protein that binds to the intronic cluster of RNA regulatory elements, downstream control sequence (DCS). It is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein but it is expressed primarily in the brain.Specifications
PTBP2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q66H20, Q91Z31, Q9UKA9 | |
310820, 56195, 58155 | |
Peptide | |
Primary | |
-20°C | |
PTBP2 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
brPTB, PTB, PTB like protein, PTBLP, PTBP2 | |
Ptbp2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |