missing translation for 'onlineSavingsMsg'
Learn More

PTBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55186-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16836675

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to PTBP2.

PTBP2 encodes a protein that binds to the intronic cluster of RNA regulatory elements, downstream control sequence (DCS). It is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein but it is expressed primarily in the brain.
Specifications

Specifications

PTBP2
Polyclonal
Unconjugated
PTBP2
brPTB, PTB, PTB like protein, PTBLP, PTBP2
Rabbit
Antigen Affinity Chromatography
RUO
310820, 56195, 58155
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q66H20, Q91Z31, Q9UKA9
Ptbp2
Peptide
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers