Learn More
Sampling Systems™ PTFE Bottles
PTFE bottles are ideal for storage and are made from 100% PTFE, giving them high chemical resistance. They are suitable for low temperatures and include both a bottle and a cap.
Brand: Sampling Systems™ A125-2000
This item is not returnable.
View return policy
Description
- Conforms to both FDA and USP class VI requirements
- Crevice-free interior
- Heavy wall construction
- Suitable for use up to 280°C
Specifications
2 L | |
White | |
Non-sterile | |
Autoclavable | |
No | |
Cylindrical | |
Storage Bottle | |
PTFE | |
Yes |
67.6 oz. | |
PTFE | |
Yes | |
Reusable | |
Wide | |
Certified Quality | |
Unlined | |
Screw Cap |
Product Content Correction
Product Title
Sampling Systems™ PTFE Bottles > PTFE Bottles; Capacity (Metric): 2 L
Spot an opportunity for improvement?Share a Content Correction