Learn More
Bohlender™ PTFE Distributors for Bottles
A black screw cap made of PPS for bottle thread GL 45, excellent for use in HPLC applications.
975.00 SEK - 1845.00 SEK
Specifications
Temperature Range (Metric) | −50° to +200°C |
---|---|
Thread Size (Metric) | GL 45 |
Closure Color | Black |
Certifications/Compliance | FDA conformity |
For Use With (Equipment) | Tubing I.D. 1.6 mm x O.D. 3.2 mm |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
10023491
|
Bohlender™
F746-04 |
2 Stopcocks |
975.00 SEK
Each |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
10678631
|
Bohlender™
F746-12 |
3 Stopcocks |
1845.00 SEK
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||||||
Description
- Providing temperature resistance from −50° to +200°C
- Universal chemical resistance
- FDA conformity
- Body made of PTFE with 2 or 3 ports with female thread UNF 0.25 in. 28 G on upper and lower sides and stopcock made of FEP for each port
- Possible unevenness of the bottle neck is adjusted by an o-ring behind an elastic sealing lip
- Body of the distributor can be turned independently from the screw cap
- Completely assembled distributor can be removed completely and fixed on another bottle without the risk of disarranging the tubing
Specifications
−50° to +200°C | |
Black | |
Tubing I.D. 1.6 mm x O.D. 3.2 mm |
GL 45 | |
FDA conformity |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bohlender™ PTFE Distributors for Bottles