Learn More
Fisherbrand™ PTFE Stopper, Flask with Handle
Fisherbrand™ PTFE Stopper, Flask with Handle are for use with volumetric flasks and similar containers.
Brand:  Fisherbrand™ 010.4.38
 View more versions of this product
Product Code. 16235971
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Solid Construction
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Specifications
Fisherbrand™ PTFE Stopper, Flask with Handle, Black Handle
1
-200°C to +280°C
PTFE
Standard Taper Ground Glass Joints
38 (US)
Black
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ PTFE Stopper, Flask with Handle > Size (US): 38; Handle Color: Black