Learn More
Fisherbrand™ PTFE Tweezers, Fine Sharp
Fisherbrand™ PTFE Tweezers, Fine Sharp are inert and non-contaminating
Brand:  Fisherbrand™ 027.0151
 View more versions of this product
Product Code. 16286021
  • 142.00 SEK / Each
Only null left
Estimated Shipment: 04-08-2025
Add to basket
Description
Description
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Specifications
PTFE
150 mm
Fisherbrand™ PTFE Tweezers, Fine Sharp
1 Ea.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ PTFE Tweezers, Fine Sharp > Length: 150 mm