Learn More
PTGER4 Mouse anti-Human, Unconjugated, Clone: 4A2A12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66921-1-IG-20UL
 View more versions of this product
Product Code. 16889215
  • 1645.54 SEK / 20µL
Description
The PTGER4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTGER4. This antibody recognizes human, mouse, rat, pig antigen. The PTGER4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
PTGER4
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
4A2A12
Unconjugated
20 μL
5734
IgG2b
Certificates
Promotions
Promotions
Product Content Correction
Product Title
PTGER4 Mouse anti-Human, Unconjugated, Clone: 4A2A12, Proteintech > 20 μL