missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN11 Mouse anti-Human, Mouse, Rat, Clone: 3F8A8, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PTPN11 |
---|---|
Clone | 3F8A8 |
Concentration | 0.8 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888995
|
Proteintech
66795-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878995
|
Proteintech
66795-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains two tandem Src homology-2 domains, which function as phospho-tyrosine binding domains and mediate the interaction of this PTP with its substrates. This PTP is widely expressed in most tissues and plays a regulatory role in various cell signaling events that are important for a diversity of cell functions, such as mitogenic activation, metabolic control, transcription regulation, and cell migration. Mutations in this gene are a cause of Noo syndrome as well as acute myeloid leukemia.Specifications
PTPN11 | |
0.8 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BPTP3, CFC, NS1, PTP 1D, PTP 2C, PTP2C, PTPN11, SH PTP2, SH PTP3, SHP 2, SHP2, SHPTP2 | |
PTPN11 | |
IgG1 | |
Protein G | |
Antibody |
3F8A8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Rat, Human | |
P35235, P41499, Q06124 | |
19247, 25622, 5781 | |
PTPN11 Fusion Protein Ag13649 | |
Primary | |
-20°C | |
PTPN11 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PTPN11 Mouse anti-Human, Mouse, Rat, Clone: 3F8A8, Proteintech