missing translation for 'onlineSavingsMsg'
Learn More

PTPN18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1706.43 SEK - 4863.32 SEK

Specifications

Antigen PTPN18
Concentration 0.32 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897454
View Documents
Proteintech
17551-1-AP-20UL
20 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16887454
View Documents
Proteintech
17551-1-AP-150UL
150 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Differentially dephosphorylate autophosphorylated tyrosine kinases which are known to be overexpressed in tumor tissues.
Specifications

Specifications

PTPN18
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Human, Rat
Q61152, Q99952
19253, 26469, 301333
PTPN18 Fusion Protein Ag11704
Primary
-20°C
PTPN18
0.32 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
BDP1, Brain derived phosphatase, PTP HSCF, PTPN18
Ptpn18
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PTPN18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.