missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12109-1-AP-150UL
This item is not returnable.
View return policy
Description
MEG2 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal domain that shares a significant similarity with yeast SEC14, which is a protein that has phosphatidylinositol transfer activity and is required for protein secretion through the Golgi complex in yeast. This PTP was found to be activated by polyphosphoinositide, and is thought to be involved in signaling events regulating phagocytosis.Specifications
PTPN9 | |
Polyclonal | |
Unconjugated | |
PTPN9 | |
MEG2, PTPase MEG2, PTPMEG2, PTPN9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
266611, 56294, 5780 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35239, P43378, Q641Z2 | |
Ptpn9 | |
PTPN9 Fusion Protein Ag2754 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PTPN9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction