Learn More
Labcon™ PurePlus™ 1.2 mL Sample Library Tubes
Sample library tubes come in strips of 8, 12, or as individual tubes.
Brand:  Labcon™ 3910-545-000-9
 View more versions of this product
Product Code. 17527936
  • 835.08 SEK / Pack of 960
Only null left
Estimated Shipment: 12-08-2025
Description
  • Available in 96-place racks or resealable bags
  • ATP, RNase/DNase, Endotoxin-free (non-pyrogenic)
Specifications
Specifications
Autoclavable
Polypropylene
960/Pk.
1.2 mL
Clear
Individual Tubes, Rack
960 Tubes/Pk., 10 Pk./Cs.
Sterile
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Labcon™ PurePlus™ 1.2 mL Sample Library Tubes > 1.2 mL Sample Library Tubes; Format: Individual Tubes, Rack; Sterility: Sterile; Volume (Metric): 12 mL; Color: Clear; Material: Polypropylene