Learn More
Buerkle™ PVC Standard Bung
Laboratory standard bung made of PVC with good chemical resistance against acids and alkalis.
Brand: Buerkle™ 9763-1115
11.09 SEK valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.00300kg
Alert:
Attention! Make sure to pay attention to the “List of Chemical Resistance“!
Description
- PVC, soft, red
Specifications
Red | |
20 mm | |
15 mm |
Standard Bung | |
11 mm | |
PVC |