Läs mer
Pyrimethanil, TRC
Produktkod. 30133899
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133899 10 g
30010812 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133899 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99725010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53112-28-0
Molekylformel C12 H13 N3
Molekylvikt (g/mol) 199.25
Synonym 2-Pyrimidinamine, 4,6-dimethyl-N-phenyl- (9CI, ACI), 4,6-Dimethyl-N-phenyl-2-pyrimidinamine (ACI), 2-Anilino-4,6-dimethylpyrimidine, Mythos, Mythos 300SC, N-(4,6-Dimethylpyrimidin-2-yl)aniline, Penbotec, Pyrimethanil, Scala, Siganex, Xedathane 20
IUPAC-namn 4,6-dimethyl-N-phenylpyrimidin-2-amine
LEDER Cc1cc(C)nc(Nc2ccccc2)n1
Specifikationer
Kemiskt namn eller material Pyrimethanil
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C12H13N3/c1-9-8-10(2)14-12(13-9)15-11-6-4-3-5-7-11/h3-8H,1-2H3,(H,13,14,15)
Formel vikt 199.1109
Procent renhet >95%