CAS RN 53112-28-0
IUPAC Namn:
4,6-dimethyl-N-phenylpyrimidin-2-amine
Synonymer:
pyrimethanil
Molekylvikt (g/mol):
199.26
Molekylär sammansättning:
C12H13N3
InChi Key:
ZLIBICFPKPWGIZ-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=NC(NC2=CC=CC=C2)=N1
1
–
5
av
5
Resultat
1
Pyrimethanil 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Pyrimethanil, TRC
CAS: 53112-28-0 Molekylformel: C12 H13 N3 Molekylvikt (g/mol): 199.25 Synonym: 2-Pyrimidinamine, 4,6-dimethyl-N-phenyl- (9CI, ACI),4,6-Dimethyl-N-phenyl-2-pyrimidinamine (ACI),2-Anilino-4,6-dimethylpyrimidine,Mythos,Mythos 300SC,N-(4,6-Dimethylpyrimidin-2-yl)aniline,Penbotec,Pyrimethanil,Scala,Siganex,Xedathane 20 IUPAC-namn: 4,6-dimethyl-N-phenylpyrimidin-2-amine LEDER: Cc1cc(C)nc(Nc2ccccc2)n1
5
Pyrimethanil, Fluka™
Analytical standard, reference material