Learn More
Thermo Scientific™ Quantech™ Sharp Cut Fluorometer Filters
Thermo Scientific™ Quantech Sharp Cut Fluorometer Filters deliver accurate results with the sensitivity you demand.
3095.70 SEK - 5768.10 SEK
Specifications
Type Sharp Cut Filter
For Use With (Equipment) Quantech
View More Specs
Products 11
Product Code Brand Wavelength Price Quantity & Availability  
Product Code Brand Wavelength Price Quantity & Availability  
11721131
View Documents
Thermo Scientific™
LE1095X18
415 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10314110
View Documents
Thermo Scientific™
LE1095X19
430 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11741131
View Documents
Thermo Scientific™
LE1095X20
450 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11751131
View Documents
Thermo Scientific™
LE1095X21
475 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11761131
View Documents
Thermo Scientific™
LE1095X22
500 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11651369
View Documents
Thermo Scientific™
LE1095X23
515 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11771131
View Documents
Thermo Scientific™
LE1095X24
535 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11781131
View Documents
Thermo Scientific™
LE1095X25
550 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11791131
View Documents
Thermo Scientific™
LE1095X26
585 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11701141
View Documents
Thermo Scientific™
LE1095X27
605 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11711141
View Documents
Thermo Scientific™
LE1095X29
665 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Thermo Scientific™ Quantech Sharp Cut Fluorometer Filters deliver accurate results with the sensitivity you demand.
Sharp cut filters allow all of the light beyond the cut off wavelength to pass through the filter - delivering the maximum amount of signal to the detector for accurate fluorescence assays.
Sharp cut filters can be used to measure fluorescence emission and increase detection limits for sensitive measurements.
To select the proper filter for your assay, use the following guidelines:
  • There is no overlap between the Narrow Band excitation filter and the Sharp Cut emission filter The majority of the fluorescence emission falls in the wavelength range transmitted by the Sharp Cut emission filter
Includes:
SC415 filter
Note:
Number following SC is wavelength in nanometers. Sharp Cut filters transmit light to the red (lower energy) of the indicated wavelength, and therefore allow the majority of the fluorescence emission to pass through the filter. Should never be used as excitation filters.
Specifications
Specifications
Sharp Cut Filter
Quantech
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Quantech™ Sharp Cut Fluorometer Filters