Learn More
Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 6538™
Description
Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 6538™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
- No serial dilution required
- Delivers less-than100 colony forming units (CFUs) in a single 0.1mL inoculum
- 10 tests/reconstituted vial; 100 tests/kit
- Ready for rehydration
- For use in quality control proceduresbullets1 (end)
Includes:
10 preserved organism vials and 10 rehydration vials.
Specifications
Specifications
ATCC® | 6538TM* |
Description | Staphylococcus aureus subsp. aureus ATCC™ 6538™ |
Format | Quanti-Cult Plus |
Product Type | Quality Control Organism |
Target Organism Class | Cocci |
Test Type | Quantitative QC |
Packaging Type | 100 Test/Kit |
Quantity | 100 Tests/Kit |
Strain Designations | FDA 209 |
Documents
Scientific Resources
Brochures
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.