Learn More
Sartorius Quantitative Grade 390 Filter Papers
These filter papers are used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration. They are made out of 100 % cotton linters with an α-cellulose content of > 98 % and are acid-washed to make the papers ashless and achieve high purity.
146.00 SEK - 3270.00 SEK
Specifications
Color Code Green
Material 100% Cotton Linters (>98%alpha-cellulose, Ash-free)
Sterility Non-sterile
Grade 390
Quantity 100/Pk.
View More Specs
Products 11
Product Code Brand Format Filtration Speed Particle Retention Diameter (Metric) Price Quantity & Availability  
Product Code Brand Format Filtration Speed Particle Retention Diameter (Metric) Price Quantity & Availability  
10016861
View Documents
Sartorius
FT-3-103-110
Disc 100 sec. 3 to 5 μm 110 mm
421.00 SEK
Pack of 100
Only null left
In Stock
Add to basket
10541922
View Documents
Sartorius
FT-3-103-125
Disc 100 sec. 3 to 5 μm 125 mm
494.00 SEK
Pack of 100
Only null left
Estimated Shipment: 25-08-2025
Add to basket
10042431
View Documents
Sartorius
FT-3-103-185
Disc 100 sec. 3 to 5 μm 185 mm
1065.00 SEK
Pack of 100
Only null left
In Stock
Add to basket
10551922
View Documents
Sartorius
FT-3-103-150
Disc 100 sec. 3 to 5 μm 150 mm
710.00 SEK
Pack of 100
Only null left
In Stock
Add to basket
11711385
View Documents
Sartorius
FT-4-103-150
Folded 100 sec. 3 to 5 μm 125 mm
985.00 SEK
Pack of 100
Only null left
Estimated Shipment: 03-09-2025
Add to basket
10575891
View Documents
Sartorius
FT-3-103-055
Disc 100 sec. 3 to 5 μm 55 mm
146.00 SEK
Pack of 100
Only null left
In Stock
Add to basket
10092401
View Documents
Sartorius
FT-3-103-090
Disc 100 sec. 3 to 5 μm 90 mm
306.00 SEK
Pack of 100
Only null left
In Stock
Aggiungi al carrello
10624522
missing translation for 'viewDocuments'
Sartorius
FT-3-103-070
Disc 100 sec. 3 to 5 μm 70 mm
191.00 SEK
Pack of 100
Solo null disponibili
In Stock
Aggiungi al carrello
11701385
missing translation for 'viewDocuments'
Sartorius
FT-4-103-125
Folded 250 sec. 1 to 2 μm 125 mm
913.00 SEK
Pack of 100
Solo null disponibili
Estimated Shipment: 03-09-2025
Aggiungi al carrello
15932980
missing translation for 'viewDocuments'
Sartorius
FT-4-103-185
Folded 100 sec. 3 to 5 μm 185 mm
1210.00 SEK
Pack of 100
Solo null disponibili
Estimated Shipment: 08-09-2025
Aggiungi al carrello
10576081
missing translation for 'viewDocuments'
Sartorius
FT-3-103-240
Disc 100 sec. 3 to 5 μm 240 mm
3270.00 SEK
Pack of 100
Solo null disponibili
Estimated Shipment: 22-08-2025
Aggiungi al carrello
Descrizione
Descrizione
  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifica
Specifica
Green
Non-sterile
100/Pk.
100% Cotton Linters (>98%alpha-cellulose, Ash-free)
390
Quantitative Filter Paper
Videos
SDS
Documenti
Documenti
Certificati
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Sartorius Quantitative Grade 390 Filter Papers
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato