missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quercetin dihydrate, 99.85%, For HPLC analysis, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
Specifications
Specifications
Color | Green-Yellow |
Quantity | 10 g |
Formula Weight | 338.26 |
Percent Purity | 0.9985 |
Physical Form | Powder |
Chemical Name or Material | Quercetin dihydrate, Molecular Biology |
Safety and Handling

Product Identifier
- Quercetin Dihydrate
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 3,3',4',5,7-Pentahydroxyflavone dihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Quercetin dihydrate, 99.85%, For HPLC analysis, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction