Learn More
SCAT Quick-Lock Connectors
Quick-Lock Connectors provide easy and safe connection of capillary tubing and SCAT Safety Caps. Connect and disconnect with a click - without dripping or air bubbles. The valve closes automatically when disconnected. Included PFA fittings allow all common hose diameters to be connected.
Brand: SCAT 360179
429.52 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The Quick-Lock Connector sets are suitable for connecting 2 capillaries (Set 1) or one capillary and one SCAT Safety Cap (Set 2).- Made of chemically resistant glass-filled polypropylene, PEEK™ materials
- Leak protection thanks to integrated shut-off valves
- Closed system for drip-free disconnection with very low air entrapment
- Minimizes contamination and no need to purge air bubbles from the system
- Suitable for capillaries with outer diameter 1.6 (1/16 in.) / 2.3 /3.2 (1/8 in.) mm
- User-friendly, ergonomic design, enables safe connection
- Up to vacuum 15 psi, 1.0 bar pressure resistance
Specifications
HPLC Systems | |
Capillaries, Tubes and Safety Caps | |
1 x PFA-fitting 1.6/2.3/3.2 mm ID | |
Quick-Lock Connector (f) |
Videos
-
HOW TO Use HPLC Quick Connectors with SCAT Safety Caps