Learn More
DWK Life Sciences Quick Release Clamp, from Stainless steel, with retaining clip
The quick release clamp is used to close flat flange vessels.
775.00 SEK - 1325.00 SEK
Spezifikation
Quantity | 1 Ea. |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Quick Release Clamp for Closure |
For Use With (Application) | Quick release clamp for closure; Accessories for flat flange vessels |
Produktcode | Marke | Size | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Size | Preis | Menge & Verfügbarkeit | |||||
10100161
|
DWK Life Sciences
290714609 |
DN 100 |
985.00 SEK
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
10464691
|
DWK Life Sciences
290713407 |
DN 60 |
775.00 SEK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10683331
|
DWK Life Sciences
290715108 |
DN 120 |
1040.00 SEK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
12656203
|
DWK Life Sciences
290715708 |
DN 150 |
1160.00 SEK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10170201
|
DWK Life Sciences
290716104 |
DN 200 |
1325.00 SEK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Beschreibung
The quick release clamp closes flat flange vessels and secures the lid. Slipping or unchecked opening is not possible. The closure is made of high-quality Stainless steel and has three holding segments.
- With retaining clip
- Quick closing with quick-release fastener
Spezifikation
1 Ea. | |
Stainless Steel | |
Quick Release Clamp for Closure | |
Quick release clamp for closure; Accessories for flat flange vessels |