Learn More
Quickfit™ Glass Stirrer Gland
Introduce stirrer paddles into Quickfit™ flasks and vessels with these glass stirrer glands. Quickfit™ Glass Stirrer Glands provide a vapor-tight seal between paddle and flask.
Brand: Quickfit ST20/3
1976.00 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.14100kg
Description
- Holds a vacuum down to 0.1 torr (13.3Pa)
- Comprises parallel ground glass sleeve with cup to accept a suitable lubricant
- Supplied complete with screwcaps rubber rings and PTFE washer
Specifications
Quickfit™ flasks and vessels | |
For introducing stirrer paddles into Quickfit™ flasks and vessels | |
Screw cap, Rubber rings and PTFE washers | |
24/29 |