Learn More
Quickfit™ Glass Stirrer Gland
Introduce stirrer paddles into Quickfit™ flasks and vessels with these glass stirrer glands. Quickfit™ Glass Stirrer Glands provide a vapor-tight seal between paddle and flask.
1500.00 SEK - 10570.00 SEK
Specifications
For Use With (Equipment) | Quickfit™ flasks and vessels |
---|---|
For Use With (Application) | For introducing stirrer paddles into Quickfit™ flasks and vessels |
Includes | Screw cap, Rubber rings and PTFE washers |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10238024
|
Quickfit
ST20/2 |
19/26 |
1500.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
12003314
|
Quickfit
ST20/3 |
24/29 |
2080.00 SEK
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
15567990
|
Quickfit
ST20/4 |
29/32 |
10570.00 SEK
Pack of 5 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
Description
- Holds a vacuum down to 0.1 torr (13.3Pa)
- Comprises parallel ground glass sleeve with cup to accept a suitable lubricant
- Supplied complete with screwcaps rubber rings and PTFE washer
Specifications
Quickfit™ flasks and vessels | |
For introducing stirrer paddles into Quickfit™ flasks and vessels | |
Screw cap, Rubber rings and PTFE washers |