missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Glass Stirrer Shaft
Designed for use with SVL™ stirrer heads and stirrer bearings
Brand: SVL 588-35
9751.75 SEK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.13000kg
Description
- Manufactured from borosilicate glass
- Lower end with internal screwthread to accept choice of stirrer heads
- Upper section of shaft is ground to fit choice of stirrer bearings
Specifications
Length: 580mm | |
SVL™ stirrer heads and stirrer bearings | |
1 Pack | |
Stirrer Shaft |