Läs mer
Quinmerac-3,8-dicarboxylic Acid (>90%), TRC
Produktkod. 30005221
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005221 50 mg
17911035 5 mg
17934715 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005221 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ69650550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90717-07-0
Molekylformel C11 H6 Cl N O4
Molekylvikt (g/mol) 251.62
Synonym Quinmerac Metabolite BH 518-2, Quinmerac metabolite BH2, 7-Chloro-3,8-quinolinedicarboxylic acid, BAS 518-2, 3,8-Quinolinedicarboxylic acid, 7-chloro-
IUPAC-namn 7-chloroquinoline-3,8-dicarboxylic acid
LEDER OC(=O)c1cnc2c(C(=O)O)c(Cl)ccc2c1
Specifikationer
Kemiskt namn eller material Quinmerac Metabolite BH 518-2
Kvantitet 50 mg
InChI formel InChI=1S/C11H6ClNO4/c12-7-2-1-5-3-6(10(14)15)4-13-9(5)8(7)11(16)17/h1-4H,(H,14,15)(H,16,17)
Formel vikt 250.9985