CAS RN 90717-07-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
Quinmerac-3,8-dicarboxylic Acid (>90%), TRC
CAS: 90717-07-0 Molekylformel: C11 H6 Cl N O4 Molekylvikt (g/mol): 251.62 Synonym: Quinmerac Metabolite BH 518-2,Quinmerac metabolite BH2,7-Chloro-3,8-quinolinedicarboxylic acid,BAS 518-2,3,8-Quinolinedicarboxylic acid, 7-chloro- IUPAC-namn: 7-chloroquinoline-3,8-dicarboxylic acid LEDER: OC(=O)c1cnc2c(C(=O)O)c(Cl)ccc2c1
2
Quinmerac metabolite BH 518-2 100 μg/mL in Acetone, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality