missing translation for 'onlineSavingsMsg'
Läs mer

(R)-2-Thioxothiazolidine-4-carboxylic Acid, TRC

Produktkod. 30137712
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137712 10 mg
30137465 50 mg
30003750 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30137712 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT39500010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 98169-56-3
Molekylformel C4 H5 N O2 S2
Molekylvikt (g/mol) 163.22
Synonym 4-Thiazolidinecarboxylic acid, 2-thioxo-, (4R)-, 4-Thiazolidinecarboxylic acid, 2-thioxo-, (R)-, (4R)-2-Thioxo-4-thiazolidinecarboxylic acid, (R)-(-)-Thiazolidine-2-thione-4-carboxylic acid, (R)-(-)-Thiazolidine-2-thione-4-carboxylic acid, (R)-Thiazolidine-2-thione-4-carboxylic acid, Raphanusamic acid
IUPAC-namn (4R)-2-thioxothiazolidine-4-carboxylic acid
LEDER OC(=O)[C@@H]1CSC(=S)N1

Specifikationer

Kemiskt namn eller material (R)-Thiazolidine-2-thione-4-carboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C4H5NO2S2/c6-3(7)2-1-9-4(8)5-2/h2H,1H2,(H,5,8)(H,6,7)/t2-/m0/s1
Formel vikt 162.9762
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.