missing translation for 'onlineSavingsMsg'
Läs mer

(R)-3-Hydroxy Myristic Acid, TRC

Produktkod. 30137246
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137246 100 mg
30137144 50 mg
30008833 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30137246 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH948505100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28715-21-1
Molekylformel C14 H28 O3
Molekylvikt (g/mol) 244.37
IUPAC-namn (3R)-3-hydroxytetradecanoic acid
LEDER CCCCCCCCCCC[C@@H](O)CC(=O)O

Specifications

Kemiskt namn eller material (R)-3-Hydroxymyristic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C14H28O3/c1-2-3-4-5-6-7-8-9-10-11-13(15)12-14(16)17/h13,15H,2-12H2,1H3,(H,16,17)/t13-/m1/s1
Formel vikt 244.2038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.