missing translation for 'onlineSavingsMsg'
Learn More
(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, 98% Product Code.: 15482456

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, 98%

Product Code. 15482456
5 mL, 5mL
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
This item is not returnable. View return policy

Product Code. 15482456

Brand: Thermo Scientific Alfa Aesar H56077.AA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109838-85-9 | C9H16N2O2 | 184.239 g/mol

(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazineChiral auxiliary for the synthesis of α-amino acids. It is an inhibitor with antitumor activity.

Solubility
Soluble in Chloroform and Ethanol

Notes
Store at -20°C. Keep away from oxidizing agents. Hygroscopic. Keep under inert atmosphere.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109838-85-9
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.239
MDL Number MFCD00040565
InChI Key FCFWEOGTZZPCTO-MRVPVSSYSA-N
Synonym r-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine, r-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, pyrazine, 2,5-dihydro-3,6-dimethoxy-2-1-methylethyl-, 2r, r-2,5-dimethoxy-3-isopropyl-3,6-dihydropyrazine, 2r-2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine, 2r-2-isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, r-2,5-dihydro-3,6-dimethoxy-2-isopropylpiperazine, 2r-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine, 2r---2,5-dihydro-3,6-dimethoxy-2-isopropylpyrazine
PubChem CID 736066
IUPAC Name (2R)-3,6-dimethoxy-2-propan-2-yl-2,5-dihydropyrazine
SMILES CC(C)C1C(=NCC(=N1)OC)OC

Specifications

Density 1.028
Flash Point 85°C (185°F)
Quantity 5 mL
Beilstein 3542790
Solubility Information Soluble in Chloroform and Ethanol
Optical Rotation −102° (c=1 in Ethanol)
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material (R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.