Learn More
R&D Systems™ Human MCHR2 (NP_115892) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Human MCHR2 (NP_115892) VersaClone cDNA clone contains the complete ORF for MCHR2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MCHR2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
Gene | mMCHR2 |
Accession Number | NP_115892 |
Concentration | 10 μg at 0.2 μg/μL |
Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
Insert Type | 1036bp |
Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Species | Mouse |
Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Product Content Correction
Product Title
R&D Systems™ Human MCHR2 (NP_115892) VersaClone cDNA >
Spot an opportunity for improvement?Share a Content Correction