Läs mer
(R)-(+)-Shikonin, TRC
Produktkod. 30134794
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134794 50 mg
30012802 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134794 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS35760050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 517-89-5
Molekylformel C16 H16 O5
Molekylvikt (g/mol) 288.3
Synonym 1,4-Naphthalenedione, 5,8-dihydroxy-2-[(1R)-1-hydroxy-4-methyl-3-penten-1-yl]-, 1,4-Naphthalenedione, 5,8-dihydroxy-2-(1-hydroxy-4-methyl-3-pentenyl)-, (R)-, 1,4-Naphthalenedione, 5,8-dihydroxy-2-[(1R)-1-hydroxy-4-methyl-3-pentenyl]- (9CI), 1,4-Naphthoquinone, 5,8-dihydroxy-2-(1-hydroxy-4-methyl-3-pentenyl)-, (+)- (8CI), Shikonin (6CI), (+)-Shikonin, (R)-(+)-Shikonin, 5,8-Dihydroxy-2-[(1R)-1-hydroxy-4-methyl-3-penten-1-yl]-1,4-naphthalenedione, C.I. 75535, Isoarnebin 4, NSC 252844, Shikonin S, Shikonine, Tokyo Violet
IUPAC-namn 5,8-dihydroxy-2-[(1R)-1-hydroxy-4-methylpent-3-enyl]naphthalene-1,4-dione
LEDER CC(=CC[C@@H](O)C1=CC(=O)c2c(O)ccc(O)c2C1=O)C
Specifikationer
Kemiskt namn eller material Shikonin
Kvantitet 50 mg
InChI formel InChI=1S/C16H16O5/c1-8(2)3-4-10(17)9-7-13(20)14-11(18)5-6-12(19)15(14)16(9)21/h3,5-7,10,17-19H,4H2,1-2H3/t10-/m1/s1
Formel vikt 288.0998