Learn More
RAB11FIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18136-1-AP-150UL
 View more versions of this product
Product Code. 16828184
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
RAB11FIP2 is an adapter protein that plays a role in the secretory pathway. It is thought to be important for endosome recycling and receptor-mediated endocytosis. In endosome recycling, RAB11-FIP2 regulates vesicle transport from the endosomal recycling compartment (ERC) to the plasma membrane.
Specifications
Specifications
RAB11FIP2
Polyclonal
Unconjugated
RAB11FIP2
KIAA0941, nRip11, Rab11 FIP2, RAB11FIP2
Rabbit
Antigen Affinity Chromatography
RUO
22841, 308003, 74998
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.79 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
G3XA57, Q7L804
Rab11fip2
RAB11FIP2 Fusion Protein Ag12736
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RAB11FIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated