missing translation for 'onlineSavingsMsg'
Learn More

RAB22A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16181-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16885714

  • 3665.00 SEK / 150µL
Estimated Shipment: 24-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody can specifically bind RAB22a.

The protein encoded by this gene is a member of the RAB family of small GTPases. The GTP-bound form of the encoded protein has been shown to interact with early-endosomal antigen 1.
Specifications

Specifications

RAB22A
Polyclonal
Unconjugated
RAB22A
Rab 22, RAB22, RAB22A, Ras related protein Rab 22A
Rabbit
Antigen Affinity Chromatography
RUO
19334, 57403
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35285, Q9UL26
RAB22A
Peptide
150 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers