Learn More
RAB27B Mouse anti-Human, Unconjugated, Clone: 2B9G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66944-1-IG-150UL
Description
The RAB27B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB27B. This antibody recognizes human antigen. The RAB27B antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
RAB27B | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2B9G9 | |
Unconjugated | |
150 μL | |
5874 | |
IgG2a |