missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB39B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12162-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the Rab family of proteins. Rab proteins are small GTPases that are involved in vesicular trafficking.Specifications
RAB39B | |
Polyclonal | |
Unconjugated | |
RAB39B | |
RAB39B, Ras related protein Rab 39B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116442, 67790 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BHC1, Q96DA2 | |
Rab39b | |
RAB39B Fusion Protein Ag2803 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RAB39B Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction