Learn More
RAB6A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10187-2-AP-20UL
 View more versions of this product
Product Code. 16826403
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Rab proteins are also an integral part of endocytic pathways. Rab 6A is a ubiquitously expressed member of the Rab family of proteins and localizes to the Golgi membrane where it regulates retrograde transport from the late endosomes via the Golgi to endoplasmic reticulum (ER) pathway. Three isoforms exist due to alternative splicing events, namely the ubiquitously expressed isoforms Rab 6A′ and Rab 6A, and the brain-specific isoform Rab 6B.
Specifications
Specifications
RAB6A
Polyclonal
Unconjugated
RAB6A
Rab 6, RAB6, RAB6A, RAB6A′, RAB6B, Ras related protein Rab 6A
Rabbit
Antigen Affinity Chromatography
RUO
19346, 5870
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P20340, P35279
RAB6A
RAB6A Fusion Protein Ag0240
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RAB6A Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated