missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABAC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10542-1-AP-150UL
This item is not returnable.
View return policy
Description
General Rab protein regulator required for vesicle formation from the Golgi complex. May control vesicle docking and fusion by mediating the action of Rab GTPases to the SNARE complexes. In addition it inhibits the removal of Rab GTPases from the membrane by GDI.Specifications
RABAC1 | |
Polyclonal | |
Unconjugated | |
RABAC1 | |
PRA1, PRA1 family protein 1, PRAF1, Rab acceptor 1 (prenylated), RABAC1, YIP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10567, 14470, 83583 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35394, Q9UI14, Q9Z0S9 | |
RABAC1 | |
RABAC1 Fusion Protein Ag0830 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RABAC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction