Learn More
ALYREF, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ALYREF.
Brand: Abnova PAB21240.100uL
Description
The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins. [provided by RefSeq
Sequence: EFGTLKKAAVHYDRSGRSLGTADVHFERKADALKAMKQYNGVPLDGRPMNIQLVTSQIDAQRRPAQSVNRGGMTRNRGAGGFGGGGGTRRGTRGGARGRGRGAGRNSKQQLSAEELDAQLDAYNARMDSpecifications
ALYREF | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant ALYREF. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q86V81 | |
ALYREF | |
Recombinant protein corresponding to amino acids of human ALYREF. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
ALYREF | |
ALY/ALY/REF/BEF/REF/THOC4 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
10189 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only