missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC102B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CCDC102B.
Brand: Abnova PAB23675.100uL
This item is not returnable.
View return policy
Description
Sequence: DSLKLSEEMKPNLDGVDLFNNGGSGNGETKTGLRLKAINLPLENEVTEISALQVHLDEFQKILWKEREMRTALEKEIERLESALSLWKWKYSpecifications
CCDC102B | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant CCDC102B. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q68D86 | |
CCDC102B | |
Recombinant protein corresponding to amino acids of human CCDC102B. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
CCDC102B | |
ACY1L/AN/C18orf14/DKFZp434K1426/DKFZp686I08254/FLJ23594/HsT1731/MGC161726/MGC161728 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
79839 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CCDC102B, Rabbit, Polyclonal Antibody, Abnova™ > 100μL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction