missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAAG1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant KAAG1.
Brand: Abnova PAB22702.100uL
This item is not returnable.
View return policy
Description
Sequence: MDDDAAPRVEGVPVAVHKHALHDGLRQVAGPGAAAAHLPRWPPPQLAASRREAPPLSQRPHRTQGAGSPPETNEKLTNPQVKESpecifications
KAAG1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant KAAG1. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
MGC78738/RU2AS | |
Rabbit | |
Antigen affinity purification | |
RUO | |
353219 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
KAAG1 | |
KAAG1 | |
Recombinant protein corresponding to amino acids of human KAAG1. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KAAG1, Rabbit, Polyclonal Antibody, Abnova™ > 100μL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction