missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metallothionein, Rabbit, Clone: N11-G, Abnova™
Rabbit monoclonal antibody raised against synthetic peptide of MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A.
Brand: Abnova MAB9787.50uL
This item is not returnable.
View return policy
Specifications
MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A | |
Monoclonal | |
Unconjugated | |
Western Blot (1:1000) ELISA (1:5000-1:10000) The optimal working dilution should be determined by the end user. | |
P04731 (Gene ID : 4489);P07438 (Gene ID : 4490);P04732 (Gene ID : 4493);P04733 (Gene ID : 4494);P13640 (Gene ID : 4495);P80294 (Gene ID : 4496);P80295 (Gene ID : 4496);Q8N339 (Gene ID : 4499);Q86YX0 (Gene ID : 4490);Q86YX5 (Gene ID : 4489);Q8TDC4 (Gene ID | |
MT1A | |
A synthetic peptide corresponding to N-terminus of human MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A. | |
RUO | |
4489/4490/4493/4494/4495/4496/644314/4500/4499/450 | |
Store at -20°C. |
ELISA, Immunoprecipitation, Western Blot | |
N11-G | |
Rabbit monoclonal antibody raised against synthetic peptide of MT1A, MT1B, MT1E, MT1F, MT1G, MT1H, MT1IP, MT1L, MT1M, MT2A. | |
In 20mM Tris-HCl, pH 8.0 (10mg/mL BSA, 0.05% sodium azide) | |
MGC32848/MT1/MT1S/MTC | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Metallothionein, Rabbit, Clone: N11-G, Abnova™ > 50μL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction