Learn More
NETO1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NETO1.
Brand: Abnova PAB25721.100ug
Description
This gene encodes a predicted transmembrane protein containing two extracellular CUB domains followed by a low-density lipoprotein class A (LDLa) domain. It also has an intracellular FXNPXY-like motif, which has been shown in other proteins to be essential for the internalization of clathrin coated pits during endocytosis. Alternatively spliced transcript variants encoding distinct proteins have been described. [provided by RefSeq
Specifications
NETO1 | |
Polyclonal | |
Unconjugated | |
Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
NETO1 | |
NETO1 | |
A synthetic peptide corresponding to 18 amino acids at C-terminus of human NETO1. | |
100 μg | |
Primary | |
NETO1 antibody is predicted to not cross-react with other NETO protein family members. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of NETO1. | |
In PBS (0.02% sodium azide) | |
BCTL1/BTCL1 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
81832 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only