Learn More
ACROBiosystems Rabbit BCMA / TNFRSF17 Protein, His Tag
Brand: ACROBiosystems BCA-R52H4-100ug
Additional Details : Weight : 0.00010kg
Description
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.5 kDa.Specifications
7.5 kDa | |
Rabbit BCMA, His Tag (BCA-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 1 - Asn 52 (Accession # G1SFN4-1). | |
-20°C | |
BCMA | |
Rabbit | |
HEK293 | |
Bio-Markers & CD Antigens |
100 μg | |
Gln 1 - Asn 52 | |
TNFRSF17,CD269,BCM,BCMA | |
Unconjugated | |
His Tag | |
Powder |