missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Rabbit BCMA / TNFRSF17 Protein, His Tag
4673.08 SEK - 34313.77 SEK
Specifications
Molecular Weight (g/mol) | 7.5 kDa |
---|---|
Source | Rabbit BCMA, His Tag (BCA-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 1 - Asn 52 (Accession # G1SFN4-1). |
Immunogen | Gln 1 - Asn 52 |
Storage Requirements | -20°C |
Gene Alias | TNFRSF17,CD269,BCM,BCMA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17216070
|
ACROBiosystems
BCA-R52H4-1MG |
1 mg |
34313.77 SEK
1mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17206070
|
ACROBiosystems
BCA-R52H4-100UG |
100 μg |
4673.08 SEK
100µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.5 kDa.Specifications
7.5 kDa | |
Gln 1 - Asn 52 | |
TNFRSF17,CD269,BCM,BCMA | |
Unconjugated | |
His Tag | |
Powder |
Rabbit BCMA, His Tag (BCA-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 1 - Asn 52 (Accession # G1SFN4-1). | |
-20°C | |
BCMA | |
Rabbit | |
HEK293 | |
Bio-Markers & CD Antigens |