missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABEP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RABEP2 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883354
|
Proteintech
14625-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873354
|
Proteintech
14625-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RABEP2, also named as RABPT5B, plays a role in membrane trafficking and in homotypic early endosome fusion. It has two isoforms with MW 59-63 kDa.Specifications
RABEP2 | |
Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q62835, Q91WG2, Q9H5N1 | |
70314, 79874, 80754 | |
RABEP2 Fusion Protein Ag6197 | |
Primary | |
-20°C | |
RABEP2 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FRA, Rabaptin 5beta, RABEP2, RABPT5B | |
Rabep2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RABEP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech