missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAC1/2/3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16117-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can recognize Rac1, Rac2, and Rac3.
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.Specifications
RAC1/2/3 | |
Polyclonal | |
Unconjugated | |
RAC1 | |
p21 Rac3, RAC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170758, 19353, 19354, 363875, 366957, 5879, 5880, 5881, 688319 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15153, P60763, P60764, P63000, P63001, Q05144, Q6RUV5 | |
Rac1, Rac2, Rac3 | |
RAC1/2/3 Fusion Protein Ag9045 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RAC1/2/3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction