missing translation for 'onlineSavingsMsg'
Learn More

RAD51AP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen RAD51AP1
Concentration 0.16 mg/mL
Applications Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16888203
View Documents
Proteintech
11255-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16878203
View Documents
Proteintech
11255-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

May participate in a common DNA damage response pathway associated with the activation of homologous recombination and double-strand break repair. Functionally cooperates with PALB2 in promoting of D-loop formation by RAD51. Binds to single and double stranded DNA, and is capable of aggregating DNA. Also binds RNA.
Specifications

Specifications

RAD51AP1
Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Rabbit
Human, Rat, Mouse
Q8C551, Q96B01
10635, 19362, 689055
RAD51AP1 Fusion Protein Ag1781
Primary
-20°C
RAD51AP1
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
PIR51, RAD51 associated protein 1, RAD51 interacting protein, RAD51AP1
RAD51AP1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RAD51AP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.