Learn More
RALA Mouse anti-Human, Unconjugated, Clone: 1H9H5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67093-1-IG-150UL
Description
The RALA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RALA. This antibody recognizes human antigen. The RALA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
RALA | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1H9H5 | |
Unconjugated | |
150 μL | |
5898 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.