Learn More
RALA Mouse anti-Human, Unconjugated, Clone: 1H9H5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67093-1-IG-150UL
 View more versions of this product
Product Code. 16879515
  • 4652.90 SEK / 150µL
Description
The RALA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RALA. This antibody recognizes human antigen. The RALA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
RALA
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1H9H5
Unconjugated
150 μL
5898
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RALA Mouse anti-Human, Unconjugated, Clone: 1H9H5, Proteintech > 150 μL